accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 35054-52-5

NOOONOOOCa

CAS RN 35054-52-5

IUPAC Name: calcium hydrate dinitrate
Synonyms: calcium hydrate dinitrate
Molecular Weight (g/mol): 182.1
Molecular Formula: CaH2N2O7
InChi Key: JJIQGEZLLWXYKV-UHFFFAOYSA-N
SMILES: O.[Ca++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Grade 
  • (6)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)