accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOH3CPOOCH3OCH3

CAS RN 35051-49-1

IUPAC Name: ethyl 2-(diethoxyphosphoryl)pentanoate
Synonyms: ethyl 2-(diethoxyphosphoryl)pentanoate
Molecular Weight (g/mol): 266.27
Molecular Formula: C11H23O5P
InChi Key: BUPVIVDUPRDDFI-UHFFFAOYNA-N
SMILES: CCCC(C(=O)OCC)P(=O)(OCC)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results