accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 35000-38-5

H3CH3CCH3OOP

CAS RN 35000-38-5

IUPAC Name: tert-butyl 2-(triphenyl-λ⁵-phosphanylidene)acetate
Synonyms: tert-butyl 2-(triphenyl-λ⁵-phosphanylidene)acetate
Molecular Weight (g/mol): 376.44
Molecular Formula: C24H25O2P
InChi Key: ZWZUFQPXYVYAFO-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (6)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)