accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 34961-64-3

OHOO

CAS RN 34961-64-3

IUPAC Name: 3-(4-formylphenyl)propanoic acid
Synonyms: 3-(4-formylphenyl)propanoic acid
Molecular Weight (g/mol): 178.19
Molecular Formula: C10H10O3
InChi Key: NRCCSDVEUWXOMG-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC=C(C=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results