accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 34941-92-9

NClF

CAS RN 34941-92-9

IUPAC Name: 4-chloro-2-fluoropyridine
Synonyms: 4-chloro-2-fluoropyridine
Molecular Weight (g/mol): 131.53
Molecular Formula: C5H3ClFN
InChi Key: GNJKJKBURMCLOR-UHFFFAOYSA-N
SMILES: FC1=NC=CC(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results