accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 349396-12-9

CH3NOOCH3

CAS RN 349396-12-9

IUPAC Name: N-benzyl-N-ethyl-4-methoxybenzamide
Synonyms: N-benzyl-N-ethyl-4-methoxybenzamide
Molecular Weight (g/mol): 269.34
Molecular Formula: C17H19NO2
InChi Key: ZEYBOROCVUQVSX-UHFFFAOYSA-N
SMILES: CCN(CC1=CC=CC=C1)C(=O)C1=CC=C(OC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results