accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 349091-96-9

CH3H3CNOOCH3

CAS RN 349091-96-9

IUPAC Name: N-benzyl-4-methoxy-N-(propan-2-yl)benzamide
Synonyms: N-benzyl-N-isopropyl-4-methoxybenzamide
Molecular Weight (g/mol): 283.37
Molecular Formula: C18H21NO2
InChi Key: MIAUCQRMTJWYDT-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)N(CC1=CC=CC=C1)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results