accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OHFFF

CAS RN 349-75-7

IUPAC Name: [3-(trifluoromethyl)phenyl]methanol
Synonyms: M-(trifluorome)benzyl alcohol
Molecular Weight (g/mol): 176.14
Molecular Formula: C8H7F3O
InChi Key: BXEHKCUWIODEDE-UHFFFAOYSA-N
SMILES: OCC1=CC=CC(=C1)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results