accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 34822-90-7

Tl

CAS RN 34822-90-7

IUPAC Name: (cyclopenta-2,4-dien-1-yl)thallane
Synonyms: cyclopentadienylthallium
Molecular Weight (g/mol): 269.48
Molecular Formula: C5H5Tl
InChi Key: RKLJDPOVAWGBSD-UHFFFAOYSA-N
SMILES: [Tl]C1C=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results