CAS RN 34737-89-8
IUPAC Name:
(3S)-1-benzyl-3-methyl-4-oxopiperidin-1-ium
Synonyms:
(3S)-1-benzyl-3-methyl-4-oxopiperidin-1-ium
Molecular Weight (g/mol):
204.29
Molecular Formula:
C13H18NO
InChi Key:
OVQAJYCAXPHYNV-NSHDSACASA-O
SMILES:
C[C@H]1C[NH+](CC2=CC=CC=C2)CCC1=O