accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 347194-02-9

H3COFFFOH

CAS RN 347194-02-9

IUPAC Name: 1-[3-(trifluoromethoxy)phenyl]ethan-1-ol
Synonyms: 1-[3-(trifluoromethoxy)phenyl]ethanol
Molecular Weight (g/mol): 206.16
Molecular Formula: C9H9F3O2
InChi Key: BWANKNAGTRWECQ-UHFFFAOYNA-N
SMILES: CC(O)C1=CC(OC(F)(F)F)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results