accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3470-54-0

IUPAC Name: 5-amino-2,3-dihydro-1H-inden-1-one
Synonyms: 5-amino-2,3-dihydroinden-1-one
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: HODOSJNSRPXYBH-UHFFFAOYSA-N
SMILES: NC1=CC=C2C(=O)CCC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results