accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H2NO

CAS RN 3470-53-9

IUPAC Name: 6-amino-1,2,3,4-tetrahydronaphthalen-1-one
Synonyms: 6-amino-3,4-dihydro-2H-naphthalen-1-one
Molecular Weight (g/mol): 161.2
Molecular Formula: C10H11NO
InChi Key: BEVVUJBVEXJGKM-UHFFFAOYSA-N
SMILES: NC1=CC=C2C(=O)CCCC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results