CAS RN 34671-83-5
IUPAC Name:
2,2'-bipyrimidine
Synonyms:
2,2'-bipyrimidine
Molecular Weight (g/mol):
158.16
Molecular Formula:
C8H6N4
InChi Key:
HKOAFLAGUQUJQG-UHFFFAOYSA-N
SMILES:
C1=CN=C(N=C1)C1=NC=CC=N1
1
–
1
of
1
results
1
2,2'-Bipyrimidine, 96%
CAS: 34671-83-5 Molecular Formula: C8H6N4 Molecular Weight (g/mol): 158.16 MDL Number: MFCD00014600 InChI Key: HKOAFLAGUQUJQG-UHFFFAOYSA-N Synonym: 2,2'-bipyrimidine,2,2'-bipyrimidyl,2,2-bipyrimidine,bpym,2-pyrimidin-2-yl pyrimidine,bipyrimidine,2,2'-bipyrimidinyl,2,2'-dipyrimidyl,2,2;-bipyrimidine PubChem CID: 123444 SMILES: C1=CN=C(N=C1)C1=NC=CC=N1