accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3460-55-7

NNHSH2N

CAS RN 3460-55-7

IUPAC Name: (4-cyanophenyl)thiourea
Synonyms: 4-cyanophenylthiourea
Molecular Weight (g/mol): 177.23
Molecular Formula: C8H7N3S
InChi Key: LFMJROANUIRGBS-UHFFFAOYSA-N
SMILES: NC(=S)NC1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results