accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 345-16-4

HOOFHO

CAS RN 345-16-4

IUPAC Name: 5-fluoro-2-hydroxybenzoic acid
Synonyms: 5-fluoro-2-hydroxybenzoic acid
Molecular Weight (g/mol): 156.11
Molecular Formula: C7H5FO3
InChi Key: JWPRICQKUNODPZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(F)=CC=C1O
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)