CAS RN 34393-59-4
IUPAC Name:
trisodium ({[5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
Synonyms:
trisodium {[5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxyphosphonate
Molecular Weight (g/mol):
469.12
Molecular Formula:
C9H12N3Na3O11P2
InChi Key:
JENUKVZGXGULDX-UHFFFAOYNA-K
SMILES:
[Na+].[Na+].[Na+].NC1=NC(=O)N(C=C1)C1OC(COP([O-])(=O)OP([O-])([O-])=O)C(O)C1O
1
–
1
of
1
results
1
Cytidine-5'-diphosphate trisodium salt, Thermo Scientific™
CAS: 34393-59-4 Molecular Formula: C9H12N3Na3O11P2 Molecular Weight (g/mol): 469.12 MDL Number: MFCD00065201 InChI Key: JENUKVZGXGULDX-LLWADOMFSA-K Synonym: cytidine 5'-diphosphate trisodium salt PubChem CID: 131674171 IUPAC Name: [(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphono hydrogen phosphate;molecular hydrogen;sodium SMILES: [Na+].[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])([O-])=O)[C@@H](O)[C@H]1O