accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 34306-42-8

H3CNHOOCH3CH3H3COHO

CAS RN 34306-42-8

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}butanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]butanoic acid
Molecular Weight (g/mol): 203.24
Molecular Formula: C9H17NO4
InChi Key: PNFVIPIQXAIUAY-UHFFFAOYNA-N
SMILES: CCC(NC(=O)OC(C)(C)C)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results