accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 343-94-2

HClH2NHN

CAS RN 343-94-2

IUPAC Name: hydrogen 2-(1H-indol-3-yl)ethan-1-amine chloride
Synonyms: hydrogen tryptamine chloride
Molecular Weight (g/mol): 196.68
Molecular Formula: C10H13ClN2
InChi Key: KDFBGNBTTMPNIG-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCCC1=CNC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)