accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3416-74-8

CH3Cl

CAS RN 3416-74-8

IUPAC Name: 1-chloronon-4-yne
Synonyms: 1-chloronon-4-yne
Molecular Weight (g/mol): 158.67
Molecular Formula: C9H15Cl
InChi Key: RHZYERSKMQJUCP-UHFFFAOYSA-N
SMILES: CCCCC#CCCCCl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)