accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3N(R)(S)OHOOHNO

CAS RN 3416-05-5

IUPAC Name: 1-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: thymidine, 3'-deoxy-
Molecular Weight (g/mol): 226.23
Molecular Formula: C10H14N2O4
InChi Key: XKKCQTLDIPIRQD-JGVFFNPUSA-N
SMILES: CC1=CN([C@H]2CC[C@@H](CO)O2)C(=O)NC1=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results