accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OONNNN

CAS RN 34114-12-0

IUPAC Name: 5-(4-carboxylatophenyl)-2H-1,2,3,4-tetrazol-2-ide
Synonyms: 5-(4-carboxylatophenyl)-2H-1,2,3,4-tetrazol-2-ide
Molecular Weight (g/mol): 188.15
Molecular Formula: C8H4N4O2
InChi Key: JMFGRJUVVYBTAX-UHFFFAOYSA-L
SMILES: [O-]C(=O)C1=CC=C(C=C1)C1=N[N-]N=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results