accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 341-58-2

IUPAC Name: 2,2'-bis(trifluoromethyl)-[1,1'-biphenyl]-4,4'-diamine
Synonyms: 2,2'-bis(trifluoromethyl)-[1,1'-biphenyl]-4,4'-diamine
Molecular Weight (g/mol): 320.24
Molecular Formula: C14H10F6N2
InChi Key: NVKGJHAQGWCWDI-UHFFFAOYSA-N
SMILES: NC1=CC(=C(C=C1)C1=C(C=C(N)C=C1)C(F)(F)F)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results