accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HOOSCl

CAS RN 3405-88-7

IUPAC Name: 2-[(4-chlorophenyl)sulfanyl]acetic acid
Synonyms: [(4-chlorophenyl)sulfanyl]acetic acid
Molecular Weight (g/mol): 202.65
Molecular Formula: C8H7ClO2S
InChi Key: YPKLXLYGMAWXDO-UHFFFAOYSA-N
SMILES: OC(=O)CSC1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results