accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3398-16-1

IUPAC Name: 4-bromo-3,5-dimethyl-1H-pyrazole
Synonyms: 4-bromo-3,5-dimethyl-1H-pyrazole
Molecular Weight (g/mol): 175.03
Molecular Formula: C5H7BrN2
InChi Key: RISOHYOEPYWKOB-UHFFFAOYSA-N
SMILES: CC1=C(Br)C(C)=NN1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)