accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3CH3HOH3C

CAS RN 33933-78-7

IUPAC Name: 5-methylnonan-5-ol
Synonyms: 5-methylnonan-5-ol
Molecular Weight (g/mol): 158.29
Molecular Formula: C10H22O
InChi Key: AGSIGVZAVLOKLP-UHFFFAOYSA-N
SMILES: CCCCC(C)(O)CCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results