accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CH3CCH3OONHOONOO

CAS RN 3392-07-2

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 2-{[(tert-butoxy)carbonyl]amino}acetate
Synonyms: 2,5-dioxopyrrolidin-1-yl 2-[(tert-butoxycarbonyl)amino]acetate
Molecular Weight (g/mol): 272.26
Molecular Formula: C11H16N2O6
InChi Key: LJCWRJYVPJJTMB-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCC(=O)ON1C(=O)CCC1=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results