accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ONBrF

CAS RN 338454-98-1

IUPAC Name: 4-[(4-bromo-2-fluorophenyl)methyl]morpholine
Synonyms: 4-[(4-bromo-2-fluorophenyl)methyl]morpholine
Molecular Weight (g/mol): 274.13
Molecular Formula: C11H13BrFNO
InChi Key: OQMLJVPTCMLEGO-UHFFFAOYSA-N
SMILES: FC1=C(CN2CCOCC2)C=CC(Br)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results