accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COINH2

CAS RN 338454-80-1

IUPAC Name: 4-iodo-2-methoxyaniline
Synonyms: 4-iodo-2-methoxyaniline
Molecular Weight (g/mol): 249.05
Molecular Formula: C7H8INO
InChi Key: AEPCMLLYVXZOLQ-UHFFFAOYSA-N
SMILES: COC1=C(N)C=CC(I)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results