CAS RN 338404-52-7
CAS RN 338404-52-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CITCO, TRC
CAS: 338404-52-7 Molecular Formula: C19H12Cl3N3OS Molecular Weight (g/mol): 436.74 Synonym: 6-(4-Chlorophenyl)imidazo[2,1-b]thiazole-5-carboxaldehyde O-[(3,4-Dichlorophenyl)methyl]oxime IUPAC Name: 1-[6-(4-chlorophenyl)imidazo[2,1-b][1,3]thiazol-5-yl]-N-[(3,4-dichlorophenyl)methoxy]methanimine SMILES: ClC1=CC=C(C2=C(/C=N/OCC3=CC=C(Cl)C(Cl)=C3)N4C(SC=C4)=N2)C=C1