accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3377-71-7

N

CAS RN 3377-71-7

IUPAC Name: 1-benzyl-1H-indole
Synonyms: 1-benzylindole
Molecular Weight (g/mol): 207.28
Molecular Formula: C15H13N
InChi Key: NJZQOCCEDXRQJM-UHFFFAOYSA-N
SMILES: C(N1C=CC2=CC=CC=C12)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results