CAS RN 33657-49-7
IUPAC Name:
chloromethyl butanoate
Synonyms:
chloromethyl butanoate
Molecular Weight (g/mol):
136.58
Molecular Formula:
C5H9ClO2
InChi Key:
BDPZFQLKFUONAG-UHFFFAOYSA-N
SMILES:
CCCC(=O)OCCl
1
–
1
of
1
results
1
Chloromethyl butyrate, 99%
CAS: 33657-49-7 Molecular Formula: C5H9ClO2 Molecular Weight (g/mol): 136.58 MDL Number: MFCD00216961 InChI Key: BDPZFQLKFUONAG-UHFFFAOYSA-N Synonym: chloromethyl butyrate,chloromethylbutyrate,butyric acid chloromethyl ester,butanoic acid, chloromethyl ester,chloromethyl n-butanoate,butyryloxymethyl chloride,acmc-1agry,ksc492g7j,butanoic acid,chloromethyl ester,n-butanoic acid chloromethyl ester PubChem CID: 2725008 IUPAC Name: chloromethyl butanoate SMILES: CCCC(=O)OCCl