accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 334827-98-4

H3CNNOH3CONNHNOH3CSOONNCH3SOOHO

CAS RN 334827-98-4

IUPAC Name: 5-{2-ethoxy-5-[(4-ethylpiperazin-1-yl)sulfonyl]pyridin-3-yl}-3-ethyl-2-(2-methoxyethyl)-2H,4H,7H-pyrazolo[4,3-d]pyrimidin-7-one; benzenesulfonic acid
Synonyms: 5-[2-ethoxy-5-(4-ethylpiperazin-1-ylsulfonyl)pyridin-3-yl]-3-ethyl-2-(2-methoxyethyl)-4H-pyrazolo[4,3-d]pyrimidin-7-one; benzenesulfonic acid
Molecular Weight (g/mol): 677.79
Molecular Formula: C29H39N7O8S2
InChi Key: STFRDYSZKVPPQF-UHFFFAOYSA-N
SMILES: OS(=O)(=O)C1=CC=CC=C1.CCOC1=NC=C(C=C1C1=NC(=O)C2=NN(CCOC)C(CC)=C2N1)S(=O)(=O)N1CCN(CC)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results