CAS RN 334521-23-2
CAS RN 334521-23-2
IUPAC Name:
(11-azidoundecyl)trimethoxysilane
Synonyms:
(11-azidoundecyl)trimethoxysilane
Molecular Weight (g/mol):
317.51
Molecular Formula:
C14H31N3O3Si
InChi Key:
ACSJXAKDVNDMME-UHFFFAOYSA-N
SMILES:
CO[Si](CCCCCCCCCCCN=[N+]=[N-])(OC)OC
1
–
1
of
1
results
1
11-Azidoundecyltrimethoxysilane, 94%
CAS: 334521-23-2 Molecular Formula: C14H31N3O3Si Molecular Weight (g/mol): 317.51 MDL Number: MFCD11982878 InChI Key: ACSJXAKDVNDMME-UHFFFAOYSA-N Synonym: 11-azidoundecyl trimethoxysilane,11-trimethoxysilyl undecyl azide PubChem CID: 46779110 SMILES: CO[Si](CCCCCCCCCCCN=[N+]=[N-])(OC)OC