accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 334017-95-7

CH3NSNH2

CAS RN 334017-95-7

IUPAC Name: 3-methylpyridine-2-carbothioamide
Synonyms: 3-methylpyridine-2-carbothioamide
Molecular Weight (g/mol): 152.22
Molecular Formula: C7H8N2S
InChi Key: PEENLLJOQZUCDN-UHFFFAOYSA-N
SMILES: CC1=C(N=CC=C1)C(N)=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results