accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 333435-04-4

H3COOOOCl

CAS RN 333435-04-4

IUPAC Name: ethyl 4-[5-(carbonochloridoyl)furan-2-yl]benzoate
Synonyms: ethyl 4-[5-(carbonochloridoyl)furan-2-yl]benzoate
Molecular Weight (g/mol): 278.69
Molecular Formula: C14H11ClO4
InChi Key: OPOPFTSILPZGEL-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(C=C1)C1=CC=C(O1)C(Cl)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results