accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OSiCH3H3CSiCH3H3COCH3

CAS RN 332343-84-7

IUPAC Name: 1,2-bis(2-methoxyphenyl)-1,1,2,2-tetramethyldisilane
Synonyms: 1,2-bis(2-methoxyphenyl)-1,1,2,2-tetramethyldisilane
Molecular Weight (g/mol): 330.57
Molecular Formula: C18H26O2Si2
InChi Key: GTVBFJHBCZGREK-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1[Si](C)(C)[Si](C)(C)C1=CC=CC=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results