accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BOClHOOH

CAS RN 332154-58-2

IUPAC Name: [3-(carbonochloridoyl)phenyl]boronic acid
Synonyms: 3-(carbonochloridoyl)phenylboronic acid
Molecular Weight (g/mol): 184.38
Molecular Formula: C7H6BClO3
InChi Key: AIHGXUQQIIYAJP-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=CC=C1)C(Cl)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results