accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 331717-63-6

CH3NNH3CNS

CAS RN 331717-63-6

IUPAC Name: 1-ethyl-3-methyl-1H-imidazol-3-ium; cyanosulfanide
Synonyms: 1-ethyl-3-methylimidazolium thiocyanate
Molecular Weight (g/mol): 169.25
Molecular Formula: C7H11N3S
InChi Key: VASPYXGQVWPGAB-UHFFFAOYSA-M
SMILES: [S-]C#N.CCN1C=C[N+](C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results