accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 331465-71-5

CH3H3CNHNNCH3CH3PNCH2H3CCH3

CAS RN 331465-71-5

IUPAC Name: 2,8,9-tris(2-methylpropyl)-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecan-5-ium
Synonyms: 2,8,9-tris(2-methylpropyl)-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecan-5-ium
Molecular Weight (g/mol): 343.52
Molecular Formula: C18H40N4P
InChi Key: WFHPXSHLCFHEIA-UHFFFAOYSA-O
SMILES: CC(C)CN1CC[NH+]2CCN(CC(C)C)P1N(CC(C)C)CC2
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results