accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CCH3H3CNH3OHO

CAS RN 33105-81-6

IUPAC Name: (2S)-2-azaniumyl-3,3-dimethylbutanoate
Synonyms: (2S)-2-ammonio-3,3-dimethylbutanoate
Molecular Weight (g/mol): 131.18
Molecular Formula: C6H13NO2
InChi Key: NPDBDJFLKKQMCM-SCSAIBSYSA-N
SMILES: CC(C)(C)[C@H]([NH3+])C([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)