accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3SOOBOHHO

CAS RN 330804-03-0

IUPAC Name: (2-methanesulfonylphenyl)boronic acid
Synonyms: 2-methanesulfonylphenylboronic acid
Molecular Weight (g/mol): 200.02
Molecular Formula: C7H9BO4S
InChi Key: MSQFFCRGQPVQRS-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC=CC=C1B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results