accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 329976-74-1

BFFFK

CAS RN 329976-74-1

IUPAC Name: potassium trifluoro(2-phenylethyl)boranuide
Synonyms: potassium trifluoro(2-phenylethyl)boranuide
Molecular Weight (g/mol): 212.06
Molecular Formula: C8H9BF3K
InChi Key: SOZVPVAYKGJPJS-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)CCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results