accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 32974-92-8

H3CNNCH3O

CAS RN 32974-92-8

IUPAC Name: 1-(3-ethylpyrazin-2-yl)ethan-1-one
Synonyms: 1-(3-ethylpyrazin-2-yl)ethanone
Molecular Weight (g/mol): 150.18
Molecular Formula: C8H10N2O
InChi Key: PPJSYGVFDJEMRP-UHFFFAOYSA-N
SMILES: CCC1=NC=CN=C1C(C)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results