accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COHCH3H3C

CAS RN 32917-52-5

IUPAC Name: 1-(2,5-dimethylphenyl)ethan-1-ol
Synonyms: 1-(2,5-dimethylphenyl)ethanol
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: VHLZFCOCNJEXTQ-UHFFFAOYNA-N
SMILES: CC(O)C1=CC(C)=CC=C1C
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)