accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3291-00-7

NH2NHO(E)

CAS RN 3291-00-7

IUPAC Name: (6E)-6-(hydrazinylmethylidene)cyclohexa-2,4-dien-1-one
Synonyms: (6E)-6-(hydrazinylmethylidene)cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 136.15
Molecular Formula: C7H8N2O
InChi Key: RHPIFAMEPPCHGX-AATRIKPKSA-N
SMILES: NN\C=C1/C=CC=CC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results