accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3290-53-7

CH3H2C

CAS RN 3290-53-7

IUPAC Name: (2-methylprop-2-en-1-yl)benzene
Synonyms: (2-methylprop-2-en-1-yl)benzene
Molecular Weight (g/mol): 132.21
Molecular Formula: C10H12
InChi Key: MXTNFIYGTWARIN-UHFFFAOYSA-N
SMILES: CC(=C)CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results