CAS RN 32890-87-2
CAS RN 32890-87-2
IUPAC Name:
2,4-bis(trifluoromethyl)benzoate
Synonyms:
2,4-bis(trifluoromethyl)benzoate
Molecular Weight (g/mol):
257.11
Molecular Formula:
C9H3F6O2
InChi Key:
JTOIZLCQNWWDCN-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C1=CC=C(C=C1C(F)(F)F)C(F)(F)F
1
–
2
of
2
results
1
Promotions Available
2,4-Bis(trifluoromethyl)benzoic acid, 98%, Thermo Scientific™
2,4-Bis(trifluoromethyl)benzoic acid, 98%, Thermo Scientific™
2
2,4-Bis(trifluoromethyl)benzoic acid, 98%, Thermo Scientific™
CAS: 32890-87-2 Molecular Formula: C9H4F6O2 Molecular Weight (g/mol): 258.119 MDL Number: MFCD00042496 InChI Key: JTOIZLCQNWWDCN-UHFFFAOYSA-N Synonym: 2,4-bis trifluoromethyl benzoic acid,2,4-bis-trifluoromethylbenzoic acid,2,4-di trifluoromethyl benzoic acid,benzoic acid, 2,4-bis trifluoromethyl,2,4-bis-trifluoromethyl-benzoic acid,maybridge4_003283,acmc-20aoeb,pubchem8491,rarechem al bo 0512 PubChem CID: 141752 IUPAC Name: 2,4-bis(trifluoromethyl)benzoic acid SMILES: C1=CC(=C(C=C1C(F)(F)F)C(F)(F)F)C(=O)O