accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 32813-50-6

SCNNO

CAS RN 32813-50-6

IUPAC Name: 4-(3-isothiocyanatopropyl)morpholine
Synonyms: 4-(3-isothiocyanatopropyl)morpholine
Molecular Weight (g/mol): 186.27
Molecular Formula: C8H14N2OS
InChi Key: BCEFDMYFAAAFPE-UHFFFAOYSA-N
SMILES: S=C=NCCCN1CCOCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results