accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

SNH2NONH2OH

CAS RN 328071-16-5

IUPAC Name: 5-(2-amino-1,3-thiazol-4-yl)-2-hydroxybenzamide
Synonyms: 5-(2-amino-1,3-thiazol-4-yl)-2-hydroxybenzamide
Molecular Weight (g/mol): 235.26
Molecular Formula: C10H9N3O2S
InChi Key: ICWYTLUIOHVZIR-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(O)C=CC(=C1)C1=CSC(N)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results